Learn More
Teknova Terrific Broth Complete with Animal-Free Soytone and Antifoam 204. 1.8L in 2L PETG, Sterile. 8 Pack.
SDP
Catalog No. 502387672
Encompass
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.
Catalog No. 50-238-7672 Supplier Teknova Supplier No. 2T87658PK
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.