Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Terrific Broth, Modified, Granulated, 500 Grams
SDP

Catalog No. 50489057
Encompass_Preferred

Enriched media for maintenance and propagation of E. Coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant problem. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.

Ingredients:

Casein Digest Peptone: 12g/l
Yeast Extract: 24g/l
Dipotassium phosphate: 9.5g/l
Monopotassium phosphate: 2.2g/

Catalog No. 50-489-057 Supplier Research Products International Corp Supplier No. T15100500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enriched media for maintenance and propagation of E. Coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant problem. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.

Ingredients:

Casein Digest Peptone: 12g/l
Yeast Extract: 24g/l
Dipotassium phosphate: 9.5g/l
Monopotassium phosphate: 2.2g/

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.