Learn More
Sigma Aldrich Fine Chemicals Biosciences Terrific Broth liqidmicr500ML
Catalog No. 502900897
Encompass
Terrific Broth is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains glycerol peptides amino acids water-soluble vitamins and carbohydrates.
Catalog No. 50-290-0897 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T5574500ML
Terrific Broth is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains glycerol peptides amino acids water-soluble vitamins and carbohydrates.