Learn More
Research Products International Corp Terrific Broth, Liquid, 1 Liter
SDP
Catalog No. 501978289
Encompass_Preferred
Sterile Filtered and Autoclaved
Terrific Broth Liquid stands as a premier growth medium meticulously crafted for pioneering research in the life sciences. Combining potent nutrients, it fosters optimal conditions for bacterial cultivation. Increases plasmid DNA yield.
Specifically developed to support higher cell densities than LB medium. 
Renowned for its unparalleled support of bacterial growth, Terrific Broth Liquid is the go-to choice for researchers seeking rapid proliferation and high yields. Its meticulously balanced formulation accommodates a diverse array of bacterial strains, making it indispensable for microbiology, molecular biology, and biotechnology applications.
Whether exploring microbial physiology, conducting genetic studies, or delving into protein expression, researchers trust Terrific Broth Liquid to catalyze breakthroughs at the forefront of life sciences research.
Catalog No. 50-197-8289 Supplier Research Products International Corp Supplier No. T881751000.0
Sterile Filtered and Autoclaved
Terrific Broth Liquid stands as a premier growth medium meticulously crafted for pioneering research in the life sciences. Combining potent nutrients, it fosters optimal conditions for bacterial cultivation. Increases plasmid DNA yield.
Specifically developed to support higher cell densities than LB medium. 
Renowned for its unparalleled support of bacterial growth, Terrific Broth Liquid is the go-to choice for researchers seeking rapid proliferation and high yields. Its meticulously balanced formulation accommodates a diverse array of bacterial strains, making it indispensable for microbiology, molecular biology, and biotechnology applications.
Whether exploring microbial physiology, conducting genetic studies, or delving into protein expression, researchers trust Terrific Broth Liquid to catalyze breakthroughs at the forefront of life sciences research.