Learn More
Alkali Scientific Terrific Broth, Liquid, 1 Liter
SDP
Catalog No. 5000660922
Encompass_Preferred
Terrific Broth, Liquid, 1 Liter, is a high-yield growth medium used in molecular biology for the cultivation of microorganisms, especially for bacterial growth at elevated rates. This liquid broth contains a high concentration of nutrients that promote fast bacterial replication, making it ideal for applications that require high-density bacterial cultures. It is frequently used in protein expression systems, such as those involving Escherichia coli, to maximize cell growth and protein production. Terrific Broth is commonly used in labs for recombinant DNA work, including plasmid propagation and gene expression experiments, making it a go-to choice for high-throughput microbiology and molecular biology applications.
Catalog No. 50-006-60922 Supplier Alkali Scientific Supplier No. TER009CP
Terrific Broth, Liquid, 1 Liter, is a high-yield growth medium used in molecular biology for the cultivation of microorganisms, especially for bacterial growth at elevated rates. This liquid broth contains a high concentration of nutrients that promote fast bacterial replication, making it ideal for applications that require high-density bacterial cultures. It is frequently used in protein expression systems, such as those involving Escherichia coli, to maximize cell growth and protein production. Terrific Broth is commonly used in labs for recombinant DNA work, including plasmid propagation and gene expression experiments, making it a go-to choice for high-throughput microbiology and molecular biology applications.