Learn More
Bioworld Terrific Broth (TB), Easy-Pack, 2 PK(s)
Catalog No. 50254453
Encompass_Preferred
Terrific Broth (TB)One step preparation of media by mixing the contents of the pack in 992 ml of H2O and 8 ml of glycerol, sterlize by autoclavingHighly enriched medium developed to improve yield in plasmid-bearing E. coliDoes not contain GlycerolpH: 7.2Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light to medium amber brothSoluble in waterContains:Bacto Tryptone Bacto Yeast Extract Dipotassium PhosphateMonopotassium Phosphate
Catalog No. 50-254-453 Supplier Bioworld Supplier No. 306210281
Terrific Broth (TB)One step preparation of media by mixing the contents of the pack in 992 ml of H2O and 8 ml of glycerol, sterlize by autoclavingHighly enriched medium developed to improve yield in plasmid-bearing E. coliDoes not contain GlycerolpH: 7.2Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light to medium amber brothSoluble in waterContains:Bacto Tryptone Bacto Yeast Extract Dipotassium PhosphateMonopotassium Phosphate