Learn More
Teknova Terrific Broth with 0.4% Glycerol. 1kg
SDP
Catalog No. 502388086
Encompass
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.
Catalog No. 50-238-8086 Supplier Teknova Supplier No. T0311
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.