Learn More
Teknova Terrific Broth with 1% Glucose and 10mM Magnesium Sulfate. 1000mL, Sterile. 8 pack.
SDP
Catalog No. 502387796
Encompass
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.
Catalog No. 50-238-7796 Supplier Teknova Supplier No. T70818PK
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.