Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Terrific Broth Modified 500ml Capsules, 200 Capsules of 24 Grams, 5 Kilograms
SDP

Catalog No. 50550443
Encompass_Preferred

Terrific Broth formulation packaged in easy to use 24 gram capsules. Formulation from powder.

Add 1 capsule per 500ml of water. Heat and stir until capsules are completely dissolved. Add 2ml of glycerol to mixture. Sterilize by autoclaving for 15 minutes.

Ingredients:

Casein Digest Peptone: 12 g/L
Yeast Extract: 24 g/L
Dipotassium Phosphate: 9.4 g/L
Potassium Phosphate: 2.2 g/L

Catalog No. 50-550-443 Supplier Research Products International Corp Supplier No. T150505000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terrific Broth formulation packaged in easy to use 24 gram capsules. Formulation from powder.

Add 1 capsule per 500ml of water. Heat and stir until capsules are completely dissolved. Add 2ml of glycerol to mixture. Sterilize by autoclaving for 15 minutes.

Ingredients:

Casein Digest Peptone: 12 g/L
Yeast Extract: 24 g/L
Dipotassium Phosphate: 9.4 g/L
Potassium Phosphate: 2.2 g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.