Learn More
TERRISS INDUSTRIES INC Certified Control Drink Flask 10 oz
Catalog No. NC0754051
Encompass
These Control Drink Flasks are designed to equate, in terms of capacity, an empty, dry container. The flasks, when properly filled, will hold the exact same content as a bottle and can be used to check proportioning.Calibrated in 1/10 or 1 ml Divisions.
Catalog No. NC0754051 Supplier TERRISS INDUSTRIES INC Supplier No. T09108
These Control Drink Flasks are designed to equate, in terms of capacity, an empty, dry container. The flasks, when properly filled, will hold the exact same content as a bottle and can be used to check proportioning.Calibrated in 1/10 or 1 ml Divisions.