Learn More
AdipoGen Territrem B (1 mg)
SDP
Supplier:  AdipoGen AGCN20142M001
Encompass_Preferred
Chemical. CAS: 70407-20-4. Formula: C29H34O9. MW: 526.6. Synthetic. Originally isolated from Aspergillus terreus. Tremorgenic mycotoxin. Potent and specific, irreversible acetylcholinesterase (AChE) inhibitor.
Catalog No. 50-205-3521