Learn More
Medchemexpress LLC Tert activator-1 | 666699-46-3 | MFCD00120536 | 99.5% | 331.77 g·mol⁻¹ | C14H12ClF2NO2S | 10 MG
SDP
Catalog No. 5000299227
Encompass_Preferred
TERT activator-1 is a small-molecule activator of telomerase reverse transcriptase used as a research reagent to study telomerase regulation, neurogenesis, and aging-related pathways. It modulates TERT transcription via the MEK/ERK/AP-1 cascade and is supplied with supporting analytical documentation for laboratory use.
  • Promotes adult neurogenesis and enhances neuromuscular function.
  • Reduces cellular senescence and systemic inflammation in aged models.
  • Small-molecule mechanism targeting TERT transcriptional activation.
  • High purity suitable for research applications.
  • Available as powder and as a DMSO solution for convenience.
  • Analytical documents available: COA, NMR, HPLC, LCMS, SDS.
Catalog No. 50-002-99227 Supplier Medchemexpress LLC Supplier No. HY40236110MG
TERT activator-1 is a small-molecule activator of telomerase reverse transcriptase used as a research reagent to study telomerase regulation, neurogenesis, and aging-related pathways. It modulates TERT transcription via the MEK/ERK/AP-1 cascade and is supplied with supporting analytical documentation for laboratory use.
  • Promotes adult neurogenesis and enhances neuromuscular function.
  • Reduces cellular senescence and systemic inflammation in aged models.
  • Small-molecule mechanism targeting TERT transcriptional activation.
  • High purity suitable for research applications.
  • Available as powder and as a DMSO solution for convenience.
  • Analytical documents available: COA, NMR, HPLC, LCMS, SDS.