Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Amyl methyl ether, MilliporeSigma™ Supelco™

Catalog No. 111020603 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0603 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0603 Supplier MilliporeSigma Supelco Supplier No. 442794
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Amyl methyl ether (TAME) is a fuel oxygenate which is utilized as an additive in gasoline to enhance the octane rating and to improve combustion.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 994-05-8
Density 0.76-0.78 g/mL (at 25°C)
Flash Point -7°C
Physical Form Neat
Linear Formula C2H5 C(CH3)2 OCH3
MDL Number MFCD00010186
Boiling Point 85°C to 86°C
Synonym 1,1-Dimethylpropyl methyl ether; Methyl 2-methyl-2-butyl ether; Methyl tert-pentyl ether; TAME
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 102.17
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.