Learn More
Sigma Aldrich Fine Chemicals Biosciences tert-Butanol puriss. p.a.500ML
Catalog No. 502898192
Encompass
tert-Butanol is a tertiary alcohol. Aqueous potassium peroxodisulphate undergoes photolysis in the presence of tert-butanol on irradiation with 254nm light at 20C. It participates in the 12-tungstophosphoric acid supported on zirconia (TPA/ZrO2) catalyzed butylation of p-cresol.
Catalog No. 50-289-8192 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 19460500ML
tert-Butanol is a tertiary alcohol. Aqueous potassium peroxodisulphate undergoes photolysis in the presence of tert-butanol on irradiation with 254nm light at 20C. It participates in the 12-tungstophosphoric acid supported on zirconia (TPA/ZrO2) catalyzed butylation of p-cresol.