Learn More
Sigma Aldrich tert-Butoxy bis(dimethylamino)methane
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Reactant for:
- Preparation of pyrroloquinazolines as photochemotherapeutic agents
- Enanatioslective formal synthesis of the cinchona alkaloid quinine via stereoselective intermolecular radical addition
- Synthesis of camptothecin via intramolecular isomuenchnone cycloaddition reaction
- Aminomethylenation reactionsreview; mild condensation reactions
Specifications
Specifications
| Density | 0.844 g/mL (at 25°C (literature)) |
| Boiling Point | 50°C to 55°C (15 mmHg) |
| Molecular Formula | C9H22N2O |
| Refractive Index | n20/D 1.422 (literature); n20/D 1.422 |
| Linear Formula | (CH3)3 COCH[N(CH3)2]2 |
| MDL Number | MFCD00042858 |
| Quantity | 10 mL |
| Synonym | Brederecks reagent |
| Molecular Weight (g/mol) | 174.28 |
| Biological Activity | Respiratory System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.