Learn More
Medchemexpress LLC [(tert-butoxycarbonyl)amino](3-chlorophenyl)acetic acid | 669713-92-2 | 98.3% | C13H16ClNO4 | 5 G
SDP
Catalog No. 5000434910
Encompass_Preferred
This product is an amino acid derivative, specifically a Glycine derivative, appearing as a white to off-white solid. Amino acid derivatives are recognized for their commercial use as ergogenic supplements, impacting the secretion of anabolic hormones, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage.
  • Appears as a white to off-white solid
  • Has a molecular weight of 285.73
  • Chemical formula is C13H16ClNO4
  • Intended for research use only
Catalog No. 50-004-34910 Supplier Medchemexpress LLC Supplier No. HY781055G
This product is an amino acid derivative, specifically a Glycine derivative, appearing as a white to off-white solid. Amino acid derivatives are recognized for their commercial use as ergogenic supplements, impacting the secretion of anabolic hormones, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage.
  • Appears as a white to off-white solid
  • Has a molecular weight of 285.73
  • Chemical formula is C13H16ClNO4
  • Intended for research use only