Learn More
Medchemexpress LLC (tert-butoxycarbonyl)glycyl-glycyl-glycyl-glycyl-glycine | 330807-97-1 | MFCD32689619 | 95.0% | 403.39 | C15H25N5O8 | 10 MG
SDP
Catalog No. 5000298554
Encompass_Preferred
Boc-gly-gly-gly-gly-gly is a Boc-protected pentaglycine peptide (tert-butoxycarbonyl glycyl pentamer) used as a protease-cleavable linker and peptide building block for research and antibody-drug conjugate development. It is supplied as a solid with CAS 330807-97-1 and molecular weight 403.39, typically provided in mg-scale pack sizes.
  • Boc-protected pentaglycine peptide, suitable for peptide synthesis.
  • Function as a protease-cleavable linker for antibody-drug conjugate design.
  • Solid form, easy to handle and store.
  • Typical reported purity ~95% (vendor-specific; check batch certificate).
  • Storage: powder stable at -20°C; in solution, store at -80°C for long-term.
  • Available in mg-scale quantities for research use.
Catalog No. 50-002-98554 Supplier Medchemexpress LLC Supplier No. HY4437010MG
Boc-gly-gly-gly-gly-gly is a Boc-protected pentaglycine peptide (tert-butoxycarbonyl glycyl pentamer) used as a protease-cleavable linker and peptide building block for research and antibody-drug conjugate development. It is supplied as a solid with CAS 330807-97-1 and molecular weight 403.39, typically provided in mg-scale pack sizes.
  • Boc-protected pentaglycine peptide, suitable for peptide synthesis.
  • Function as a protease-cleavable linker for antibody-drug conjugate design.
  • Solid form, easy to handle and store.
  • Typical reported purity ~95% (vendor-specific; check batch certificate).
  • Storage: powder stable at -20°C; in solution, store at -80°C for long-term.
  • Available in mg-scale quantities for research use.