Learn More
Medchemexpress LLC Tert-butoxycarbonyl-L-leucylglycine | 32991-17-6 | MFCD00076954 | 97.0% | 288.34 | C13H24N2O5 | 500 MG
SDP
Catalog No. 5000301923
Encompass_Preferred
(tert-Butoxycarbonyl)-L-leucylglycine is a Boc-protected dipeptide used as a building block in peptide synthesis and related organic chemistry research. It is supplied as a solid with formula C13H24N2O5 and molecular weight 288.34 g·mol-1, and is used to protect the N-terminus of leucyl residues during stepwise peptide assembly and synthetic method development.
  • Boc-protected dipeptide compatible with standard peptide coupling chemistries.
  • Solid form suitable for routine laboratory storage and handling.
  • Chemical formula C13H24N2O5 and molecular weight 288.34 g·mol-1 for exact stoichiometric calculations.
  • Suitable for incorporation into peptide chains and protecting-group strategies.
  • Provided in small-scale quantities intended for research and development use.
Catalog No. 50-003-01923 Supplier Medchemexpress LLC Supplier No. HYW039759500MG
(tert-Butoxycarbonyl)-L-leucylglycine is a Boc-protected dipeptide used as a building block in peptide synthesis and related organic chemistry research. It is supplied as a solid with formula C13H24N2O5 and molecular weight 288.34 g·mol-1, and is used to protect the N-terminus of leucyl residues during stepwise peptide assembly and synthetic method development.
  • Boc-protected dipeptide compatible with standard peptide coupling chemistries.
  • Solid form suitable for routine laboratory storage and handling.
  • Chemical formula C13H24N2O5 and molecular weight 288.34 g·mol-1 for exact stoichiometric calculations.
  • Suitable for incorporation into peptide chains and protecting-group strategies.
  • Provided in small-scale quantities intended for research and development use.