Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (tert-butoxycarbonyl)-L-valyl-L-valine | 69209-73-0 | 97.0% | 316.39 | C15H28N2O5 | 100 G
SDP

Catalog No. 5000210563
Encompass_Preferred

(tert-Butoxycarbonyl)-L-valyl-L-valine is a Boc-protected dipeptide derivative of L-valine used as an intermediate in peptide synthesis and organic chemistry. It is supplied as a white to off-white solid for research use.

  • Purity 97.0%.
  • Molecular weight 316.39 g/mol.
  • Chemical formula C15H28N2O5.
  • Appearance white to off-white solid.
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in laboratory quantities including 100 G.

Catalog No. 50-002-10563 Supplier Medchemexpress LLC Supplier No. HYW048831100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(tert-Butoxycarbonyl)-L-valyl-L-valine is a Boc-protected dipeptide derivative of L-valine used as an intermediate in peptide synthesis and organic chemistry. It is supplied as a white to off-white solid for research use.

  • Purity 97.0%.
  • Molecular weight 316.39 g/mol.
  • Chemical formula C15H28N2O5.
  • Appearance white to off-white solid.
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in laboratory quantities including 100 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.