Learn More
Medchemexpress LLC Tert-butyl 1-bromo-3,6,9,12-tetraoxapentadecan-15-oate | 564476-32-0 | MFCD22574808 | 98.0% | 385.29 g/mol | C15H29BrO6 | 1 G
SDP
Catalog No. 5000203832
Encompass_Preferred
Br-PEG4-C2-Boc is a cleavable four-unit polyethylene glycol (PEG) linker designed for use in the synthesis of antibody-drug conjugates (ADCs). It contains a bromo-functional group and a Boc-protected terminus to enable conjugation strategies for attaching cytotoxins or other payloads to antibodies. Supplied for research use, it is offered in multiple pre-weighed sizes.
  • Cleavable four-unit PEG spacer suitable for ADC construction.
  • Bromo-functionalized terminus for alkylation or substitution reactions.
  • Boc-protected terminal group for orthogonal deprotection strategies.
  • High purity suitable for research applications.
  • Available in multiple pre-weighed sizes, with larger quantities available by quote.
  • Recommended storage conditions provided for both pure form and in solvent.
Catalog No. 50-002-03832 Supplier Medchemexpress LLC Supplier No. HY1303151G
Br-PEG4-C2-Boc is a cleavable four-unit polyethylene glycol (PEG) linker designed for use in the synthesis of antibody-drug conjugates (ADCs). It contains a bromo-functional group and a Boc-protected terminus to enable conjugation strategies for attaching cytotoxins or other payloads to antibodies. Supplied for research use, it is offered in multiple pre-weighed sizes.
  • Cleavable four-unit PEG spacer suitable for ADC construction.
  • Bromo-functionalized terminus for alkylation or substitution reactions.
  • Boc-protected terminal group for orthogonal deprotection strategies.
  • High purity suitable for research applications.
  • Available in multiple pre-weighed sizes, with larger quantities available by quote.
  • Recommended storage conditions provided for both pure form and in solvent.