Learn More
Medchemexpress LLC Tert-butyl 1-bromo-3,6,9,12-tetraoxapentadecan-15-oate | 564476-32-0 | MFCD22574808 | 98.0% | 385.29 g/mol | C15H29BrO6 | 5 G
SDP
Catalog No. 5000202468
Encompass_Preferred
Br-PEG4-C2-Boc is a cleavable 4-unit polyethylene glycol (PEG) linker containing a bromoacetamido reactive handle and a Boc-protected terminal group, used in the synthesis of antibody-drug conjugates, PROTACs, and other bioconjugates in research settings.
  • Cleavable PEG4 linker suitable for conjugation chemistry.
  • Bromo group provides a reactive handle for alkylation and coupling.
  • Boc-protected terminal group enables controlled deprotection after conjugation.
  • High purity suitable for synthetic research applications.
  • Provided as a solid in multiple pack sizes for flexible workflows.
Catalog No. 50-002-02468 Supplier Medchemexpress LLC Supplier No. HY1303155G
Br-PEG4-C2-Boc is a cleavable 4-unit polyethylene glycol (PEG) linker containing a bromoacetamido reactive handle and a Boc-protected terminal group, used in the synthesis of antibody-drug conjugates, PROTACs, and other bioconjugates in research settings.
  • Cleavable PEG4 linker suitable for conjugation chemistry.
  • Bromo group provides a reactive handle for alkylation and coupling.
  • Boc-protected terminal group enables controlled deprotection after conjugation.
  • High purity suitable for synthetic research applications.
  • Provided as a solid in multiple pack sizes for flexible workflows.