Learn More
eMolecules​ 1330766-02-3 | tert-butyl 1-(hydroxymethyl)-5-azaspiro[2.4]heptane-5-carboxylate | Pharmablock | MFCD20230654 | 227.304 | C12H21NO3 | 97 | CC(C)(C)OC(=O)N1CCC2(CC2CO)C1 | 500mg | 551241212
Supplier:  eMolecules​ 551241212
Encompass_Preferred
Ambeed | (R)-N-((S)-(2-(Dicyclohexylphosphanyl)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide | 50mg | 633660088 | A1264532 | 1595319-99-5 | 483.690 | C29H42NOPS
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool<|a>
Catalog No. NC1634488