Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate, 97%

Catalog No. AC437980010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437980010 1 g Glass bottle
AC437980050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437980010 Supplier Thermo Scientific Chemicals Supplier No. 437980010
Add to Cart

CAS: 147804-30-6 | C11H19NO3 | 213.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147804-30-6
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
MDL Number MFCD07779385
InChI Key ULSBMKGFFFMGOI-UHFFFAOYSA-N
Synonym tert-butyl 1-oxa-6-azaspiro 2.5 octane-6-carboxylate, 1-oxa-6-azaspiro 2.5 octane-6-carboxylic acid, 1,1-dimethylethyl ester, 6-boc-1-oxa-6-azaspiro 2.5 octane, tert-butyl 1-oxa-6-azaspiro 2,5 octane-6-carboxylate, n-boc-1-oxa-6-azaspiro 2.5 octane, 6-n-boc-1-oxa-6-azaspiro 2.5 octane, 6-tert-butoxycarbonyl-1-oxa-6-azaspiro 2.5 octane, tert-butyl 2-oxa-6-azaspiro 2.5 octane-6-carboxylate, 1-oxa-6-aza-spiro 2.5 octane-6-carboxylic acid tert-butyl ester, tert-butyl 6-azaspiro 2.5 octane-1-oxa-6-carboxylate
PubChem CID 22135564
IUPAC Name tert-butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2(CC1)CO2

Specifications

Melting Point 58°C to 59°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 213.28
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.