Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 1-Oxa-6-azaspiro[2.5]octane-6-carboxylate 97.0+%, TCI America™
SDP

Catalog No. B55011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55011G 1 g
B55015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55011G Supplier TCI America Supplier No. B55011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 147804-30-6
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.277
MDL Number MFCD07779385
InChI Key ULSBMKGFFFMGOI-UHFFFAOYSA-N
Synonym tert-butyl 1-oxa-6-azaspiro 2.5 octane-6-carboxylate, 1-oxa-6-azaspiro 2.5 octane-6-carboxylic acid, 1,1-dimethylethyl ester, 6-boc-1-oxa-6-azaspiro 2.5 octane, tert-butyl 1-oxa-6-azaspiro 2,5 octane-6-carboxylate, n-boc-1-oxa-6-azaspiro 2.5 octane, 6-n-boc-1-oxa-6-azaspiro 2.5 octane, 6-tert-butoxycarbonyl-1-oxa-6-azaspiro 2.5 octane, tert-butyl 2-oxa-6-azaspiro 2.5 octane-6-carboxylate, 1-oxa-6-aza-spiro 2.5 octane-6-carboxylic acid tert-butyl ester, tert-butyl 6-azaspiro 2.5 octane-1-oxa-6-carboxylate
PubChem CID 22135564
IUPAC Name tert-butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2(CC1)CO2

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 1 g
Formula Weight 213.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 1-Oxa-6-azaspiro[2.5]octane-6-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.