Learn More
Medchemexpress LLC tert-Butyl 10-amino | 10g
SDP
Catalog No. 5000210078
Encompass_Preferred
tert-Butyl (10-aminodecyl)carbamate (1-Boc-1 10-diaminodecane) can be used as a PROTAC linker in the synthesis of PROTACs and other conjugation applications tert-Butyl (10-aminodecyl)carbamate is an alkane chain with terminal amine and Boc-protected amino groups Amine group is reactive with carboxylic acids activated NHS esters carbonyls (ketone aldehyde) etc The Boc group can be deprotected under mild acidic conditions to form the free amine
Catalog No. 50-002-10078 Supplier Medchemexpress LLC Supplier No. HYW04768810G
tert-Butyl (10-aminodecyl)carbamate (1-Boc-1 10-diaminodecane) can be used as a PROTAC linker in the synthesis of PROTACs and other conjugation applications tert-Butyl (10-aminodecyl)carbamate is an alkane chain with terminal amine and Boc-protected amino groups Amine group is reactive with carboxylic acids activated NHS esters carbonyls (ketone aldehyde) etc The Boc group can be deprotected under mild acidic conditions to form the free amine