Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl (10-aminodecyl)carbamate | 216961-61-4 | MFCD02094501 | ≥97.0% | 272.43 g/mol | C15H32N2O2 | 25 G
SDP

Catalog No. 5000208844
Encompass_Preferred

tert-Butyl (10-aminodecyl)carbamate is a Boc-protected primary amine used as a synthetic building block and linker in medicinal chemistry and organic synthesis. It is supplied with documentation for safe handling and quality verification.

  • Boc-protected primary amine suitable for linker and building-block synthesis.
  • High purity for research applications.
  • Provided as a 25 g pack for laboratory use.
  • Documented safety and quality data available, including SDS and COA.
  • Molecular weight 272.43 g/mol and formula C15H32N2O2.

Catalog No. 50-002-08844 Supplier Medchemexpress LLC Supplier No. 25GHYW047688
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl (10-aminodecyl)carbamate is a Boc-protected primary amine used as a synthetic building block and linker in medicinal chemistry and organic synthesis. It is supplied with documentation for safe handling and quality verification.

  • Boc-protected primary amine suitable for linker and building-block synthesis.
  • High purity for research applications.
  • Provided as a 25 g pack for laboratory use.
  • Documented safety and quality data available, including SDS and COA.
  • Molecular weight 272.43 g/mol and formula C15H32N2O2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.