Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl 11-aminoundecanoate | 220851-29-6 | 99.0% | 5 G
SDP

Catalog No. 5000338842
Encompass_Preferred

tert-Butyl 11-aminoundecanoate is a PROTAC linker, characterized by its PEG composition. It is utilized in the synthesis of various PROTACs (Proteolysis Targeting Chimeras), which are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is intended for research use only.

  • Molecular weight: 257.41
  • Chemical formula: C15H31NO2
  • CAS number: 220851-29-6
  • Appearance: Liquid (density: 0.908±0.06 g/cm3)
  • Color: White to light brown
  • Solubility in DMSO: 100 mg/mL (needs ultrasonic)

Catalog No. 50-003-38842 Supplier Medchemexpress LLC Supplier No. HY1307155G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl 11-aminoundecanoate is a PROTAC linker, characterized by its PEG composition. It is utilized in the synthesis of various PROTACs (Proteolysis Targeting Chimeras), which are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is intended for research use only.

  • Molecular weight: 257.41
  • Chemical formula: C15H31NO2
  • CAS number: 220851-29-6
  • Appearance: Liquid (density: 0.908±0.06 g/cm3)
  • Color: White to light brown
  • Solubility in DMSO: 100 mg/mL (needs ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.