Learn More
Selleck Chemical LLC tert-Butyl 11-aminoundecanoate S1814-100mg
SDP
Catalog No. 503110923
Encompass_Preferred
tert-Butyl 11-aminoundecanoate (compound 6b) is a PROTAC linker that refers to the PEG composition tert-Butyl 11-aminoundecanoate is applicable to the synthesis of a series of PROTACs
Catalog No. 50-311-0923 Supplier Selleck Chemical LLC Supplier No. S1814100MG
tert-Butyl 11-aminoundecanoate (compound 6b) is a PROTAC linker that refers to the PEG composition tert-Butyl 11-aminoundecanoate is applicable to the synthesis of a series of PROTACs