Learn More
Medchemexpress LLC Tert-butyl (11-aminoundecyl)carbamate | 937367-26-5 | MFCD02094502 | ≥97.0% | 286.45 g/mol | C16H34N2O2 | 100 MG
SDP
Catalog No. 5000282992
Encompass_Preferred
N-Boc-undecane-1,11-diamine is a Boc-protected diamine with an 11-carbon alkyl spacer, supplied as a white to off-white solid and used as a building block in organic synthesis. It is commonly employed as a flexible linker in PROTAC assembly and conjugation workflows; the Boc protecting group can be removed under mild acidic conditions to reveal the free amine for further functionalization.
  • Boc-protected primary diamine enabling selective deprotection.
  • Eleven-carbon flexible spacer suitable for linker design.
  • Solid form for easy handling and storage.
  • High purity (≥97.0%) suitable for synthetic applications.
  • CAS number 937367-26-5 for unambiguous identification.
Catalog No. 50-002-82992 Supplier Medchemexpress LLC Supplier No. HYW131541100MG
N-Boc-undecane-1,11-diamine is a Boc-protected diamine with an 11-carbon alkyl spacer, supplied as a white to off-white solid and used as a building block in organic synthesis. It is commonly employed as a flexible linker in PROTAC assembly and conjugation workflows; the Boc protecting group can be removed under mild acidic conditions to reveal the free amine for further functionalization.
  • Boc-protected primary diamine enabling selective deprotection.
  • Eleven-carbon flexible spacer suitable for linker design.
  • Solid form for easy handling and storage.
  • High purity (≥97.0%) suitable for synthetic applications.
  • CAS number 937367-26-5 for unambiguous identification.