Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl (11-aminoundecyl)carbamate | 937367-26-5 | MFCD02094502 | >97.0% | 286.45 g/mol | C16H34N2O2 | 25 MG
SDP

Catalog No. 5000283305
Encompass_Preferred

N-Boc-undecane-1,11-diamine is a Boc-protected primary diamine with an eleven-carbon alkyl spacer commonly used as a linker building block in PROTAC synthesis and other conjugation chemistries. The Boc protecting group can be removed under mild acidic conditions to reveal a free terminal amine for subsequent functionalization.

  • Boc-protected primary diamine for linker construction.
  • Eleven-carbon alkyl spacer provides flexible separation between functional groups.
  • Purity >97.0% suitable for most research applications.
  • Molecular weight 286.45 g/mol; chemical formula C16H34N2O2.
  • Stable when stored at 4°C and protected from light; long-term storage at -80°C recommended.
  • Available in small-scale quantities for laboratory use.

Catalog No. 50-002-83305 Supplier Medchemexpress LLC Supplier No. HYW13154125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Boc-undecane-1,11-diamine is a Boc-protected primary diamine with an eleven-carbon alkyl spacer commonly used as a linker building block in PROTAC synthesis and other conjugation chemistries. The Boc protecting group can be removed under mild acidic conditions to reveal a free terminal amine for subsequent functionalization.

  • Boc-protected primary diamine for linker construction.
  • Eleven-carbon alkyl spacer provides flexible separation between functional groups.
  • Purity >97.0% suitable for most research applications.
  • Molecular weight 286.45 g/mol; chemical formula C16H34N2O2.
  • Stable when stored at 4°C and protected from light; long-term storage at -80°C recommended.
  • Available in small-scale quantities for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.