Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tert-Butyl 12-Amino-4,7,10-Trioxadodecanoate

Catalog No. 830601GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-060-1GF 5 g
83-060-5GF 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 83-060-1GF Supplier Sigma Aldrich Supplier No. 830601GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyl 12-amino-4,7,10-trioxadodecanoate is generally used as a spacer or linker in bioconjugate chemistry. Some of its applications are:Synthesis of the tetanus-toxin conjugate of MUC1 glycopeptide antigen, as a potential antitumor vaccine.Synthesis of self-assembled monolayer-based surface-enhanced raman scattering (SERS) labels for immuno-SERS microscopy.Synthesis of polycationic adamantane-based dendrons for cell imaging.Synthesis of phthalocyanine (Pc)-peptide conjugates as potential fluorescence imaging agents for cancers overexpressing epidermal growth factor receptors (EGFR).Intermediate in the synthesis of a spacer for combinatorial chemistry

Specifications

CAS 252881-74-6
Molecular Formula C13H27NO5
Linear Formula C13H27NO5
MDL Number MFCD06201017
Quantity 5 g
Synonym Amino-PEG3-tert-butyl ester; tert-Butyl 3-[2-(2-(2-aminoethoxy)ethoxy)ethoxy]propionate
Molecular Weight (g/mol) 277.36 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.