Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate 97.0+%, TCI America™
SDP

Catalog No. B55871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55871G 1 g
B55875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55871G Supplier TCI America Supplier No. B55871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 186020-66-6
Molecular Formula C13H26O6
Molecular Weight (g/mol) 278.345
MDL Number MFCD03791106
InChI Key KSXVEOLRERRELV-UHFFFAOYSA-N
Synonym 12-Hydroxy-4,7,10-trioxadodecanoic Acid tert-Butyl Ester, tert-Butyl 3-[2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy]propionate, 3-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]propionic Acid tert-Butyl Ester
PubChem CID 3625858
IUPAC Name tert-butyl 3-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]propanoate
SMILES CC(C)(C)OC(=O)CCOCCOCCOCCO

Specifications

Color Yellow
Quantity 1 g
Formula Weight 278.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.