Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl (1S,4S,5R)-5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate | 1932042-59-5 | 97.8% | 213.28 | 1 G
SDP

Catalog No. 5000404254
Encompass_Preferred

tert-Butyl (1S,4S,5R)-5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate is a drug intermediate for synthesis of various active compounds.

  • Drug intermediate for synthesis of various active compounds.
  • For research use only.

Catalog No. 50-004-04254 Supplier Medchemexpress LLC Supplier No. HY34550A1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

tert-Butyl (1S,4S,5R)-5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate is a drug intermediate for synthesis of various active compounds.

  • Drug intermediate for synthesis of various active compounds.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.