Learn More
Medchemexpress LLC Tert-butyl (1S,4S,5R)-5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate | 1932042-59-5 | 97.8% | 213.28 | 250 MG
SDP
Catalog No. 5000402693
Encompass_Preferred
This product is a drug intermediate for the synthesis of various active compounds.
  • Drug intermediate target
  • For research use only
  • Solid appearance
  • White to off-white color
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-02693 Supplier Medchemexpress LLC Supplier No. 250MGHY34550A
This product is a drug intermediate for the synthesis of various active compounds.
  • Drug intermediate target
  • For research use only
  • Solid appearance
  • White to off-white color
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month