Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl (1S,4S,5R)-5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate | 1932042-59-5 | 97.8% | 213.28 | 50 MG
SDP

Catalog No. 5000403808
Encompass_Preferred

tert-Butyl (1S,4S,5R)-5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only.

  • Molecular weight: 213.28
  • Formula: C11H19NO3
  • Appearance: Solid
  • Color: White to off-white
  • Purity (batch specific): 97.77%

Catalog No. 50-004-03808 Supplier Medchemexpress LLC Supplier No. HY34550A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl (1S,4S,5R)-5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only.

  • Molecular weight: 213.28
  • Formula: C11H19NO3
  • Appearance: Solid
  • Color: White to off-white
  • Purity (batch specific): 97.77%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.