Learn More
Medchemexpress LLC tert-Butyl((1S,5R,6S)-3-azabicyclo[3.3.0]octan-6-yl)carbamate | 1228030-08-7 | 95.0% | C12H22N2O2 | 1 MG
SDP
Catalog No. 5000433668
Encompass_Preferred
This chemical is tert-Butyl((1S,5R,6S)-3-azabicyclo[3.3.0]octan-6-yl)carbamate, identified for use as a laboratory chemical and in the manufacture of substances. It is a solid compound with a molecular weight of 226.32.
  • Stable under recommended storage conditions
  • Requires handling precautions to avoid inhalation and contact with eyes and skin
  • Should be stored in a cool, well-ventilated area, away from direct sunlight and ignition sources
  • Powder stable for 3 years at -20°C or 2 years at 4°C
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C
  • Shipping at room temperature if less than 2 weeks
Catalog No. 50-004-33668 Supplier Medchemexpress LLC Supplier No. HY766451MG
This chemical is tert-Butyl((1S,5R,6S)-3-azabicyclo[3.3.0]octan-6-yl)carbamate, identified for use as a laboratory chemical and in the manufacture of substances. It is a solid compound with a molecular weight of 226.32.
  • Stable under recommended storage conditions
  • Requires handling precautions to avoid inhalation and contact with eyes and skin
  • Should be stored in a cool, well-ventilated area, away from direct sunlight and ignition sources
  • Powder stable for 3 years at -20°C or 2 years at 4°C
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C
  • Shipping at room temperature if less than 2 weeks