Learn More
Medchemexpress LLC Tert-butyl (2-(2-(2-iodoethoxy)ethoxy)ethyl)carbamate | 1820026-89-8 | MFCD31580051 | >98.0% | 359.2 g·mol⁻¹ | C11H22INO4 | 250 MG
SDP
Catalog No. 5000280963
Encompass_Preferred
Tert-butyl (2-(2-(2-iodoethoxy)ethoxy)ethyl)carbamate is a Boc-protected PEG-type linker with a terminal iodine substituent. It is used as a protected amine building block and as an iodo-terminated linker for introducing functional groups via nucleophilic substitution or cross-coupling in organic synthesis and medicinal chemistry workflows.
  • Boc-protected amine for selective deprotection under mild acidic conditions.
  • Terminal iodine enables nucleophilic substitution and cross-coupling reactions.
  • PEG-type spacer improves solubility and provides controlled linker length.
  • High purity suitable for research and synthetic applications.
  • Available in small mg-scale quantities for laboratory synthesis.
Catalog No. 50-002-80963 Supplier Medchemexpress LLC Supplier No. HYW460133250MG
Tert-butyl (2-(2-(2-iodoethoxy)ethoxy)ethyl)carbamate is a Boc-protected PEG-type linker with a terminal iodine substituent. It is used as a protected amine building block and as an iodo-terminated linker for introducing functional groups via nucleophilic substitution or cross-coupling in organic synthesis and medicinal chemistry workflows.
  • Boc-protected amine for selective deprotection under mild acidic conditions.
  • Terminal iodine enables nucleophilic substitution and cross-coupling reactions.
  • PEG-type spacer improves solubility and provides controlled linker length.
  • High purity suitable for research and synthetic applications.
  • Available in small mg-scale quantities for laboratory synthesis.