Learn More
Medchemexpress LLC Tert-butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate | 1894533-96-0 | MFCD31718688 | 97.0% | 227.30 g·mol⁻¹ | C12H21NO3 | 1 G
SDP
Catalog No. 5000281486
Encompass_Preferred
tert-Butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate (CAS 1894533-96-0) is a tert-butyl-protected oxopiperidine derivative used as a building block and drug intermediate in medicinal chemistry research. It is provided in laboratory quantities for use in small-scale synthesis, with characterized purity and recommended storage conditions for long-term stability.
  • Used as a drug intermediate in small-scale synthesis.
  • Provides tert-butyl protection for piperidinone derivatives.
  • Molecular formula C12H21NO3; molecular weight 227.30 g·mol⁻¹.
  • Typical supplied purity is about 97.0%.
  • Stable at room temperature for long-term storage when dry.
Catalog No. 50-002-81486 Supplier Medchemexpress LLC Supplier No. HYW4517561G
tert-Butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate (CAS 1894533-96-0) is a tert-butyl-protected oxopiperidine derivative used as a building block and drug intermediate in medicinal chemistry research. It is provided in laboratory quantities for use in small-scale synthesis, with characterized purity and recommended storage conditions for long-term stability.
  • Used as a drug intermediate in small-scale synthesis.
  • Provides tert-butyl protection for piperidinone derivatives.
  • Molecular formula C12H21NO3; molecular weight 227.30 g·mol⁻¹.
  • Typical supplied purity is about 97.0%.
  • Stable at room temperature for long-term storage when dry.