Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate | 1894533-96-0 | MFCD31718688 | 97.2% | 227.3 g·mol⁻¹ | C12H21NO3 | 100 MG
SDP

Catalog No. 5000281485
Encompass_Preferred

Tert-butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate is a Boc-protected piperidinone intermediate used in organic and medicinal chemistry. It is commonly employed as a building block for the synthesis of substituted piperidine derivatives and other active compounds, and is supplied for small-scale research and synthetic applications.

  • Boc-protected piperidinone intermediate.
  • High purity suitable for synthetic applications.
  • Purity approximately 97.2%.
  • Molecular weight 227.3 g·mol⁻¹.
  • Stable at room temperature with extended shelf life.
  • Suitable for medicinal chemistry and API precursor synthesis.

Catalog No. 50-002-81485 Supplier Medchemexpress LLC Supplier No. HYW451756100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tert-butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate is a Boc-protected piperidinone intermediate used in organic and medicinal chemistry. It is commonly employed as a building block for the synthesis of substituted piperidine derivatives and other active compounds, and is supplied for small-scale research and synthetic applications.

  • Boc-protected piperidinone intermediate.
  • High purity suitable for synthetic applications.
  • Purity approximately 97.2%.
  • Molecular weight 227.3 g·mol⁻¹.
  • Stable at room temperature with extended shelf life.
  • Suitable for medicinal chemistry and API precursor synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.