Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate 98.0+%, TCI America™
SDP

Catalog No. B37025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37025G 5 g
B37021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37025G Supplier TCI America Supplier No. B37025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 125995-13-3
Molecular Formula C14H28NO4
Molecular Weight (g/mol) 274.38
MDL Number MFCD07369252
InChI Key HWSHVKNLMBMKSR-GHMZBOCLSA-O
Synonym 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, tert-butyl 2-4r,6r-6-2-aminoethyl-2,2-dimethyl-1,3-dioxan-4-yl acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,cis-1,1-dimethylethyl-6-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3, 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4acetate, +/--cis-1,1-dimethylethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r-2,2-dimethyl-6beta-2-aminoethyl-1,3-dioxane-4beta-acetic acid tert-butyl ester, 4r-cis-1,1-dimethyethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate
PubChem CID 2734288
IUPAC Name 2-[(4R,6R)-6-[2-(tert-butoxy)-2-oxoethyl]-2,2-dimethyl-1,3-dioxan-4-yl]ethan-1-aminium
SMILES CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC[NH3+])OC(C)(C)O1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 273.37
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.