Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 2,6-Dichloroisonicotinate 98.0+%, TCI America™
SDP

Catalog No. B35955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35955G Supplier TCI America Supplier No. B35955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity laboratory reagent with a minimum purity of 98.0% as determined by GC analysis.

Chemical Identifiers

CAS 75308-46-2
Molecular Formula C10H11Cl2NO2
Molecular Weight (g/mol) 248.10
MDL Number MFCD08445655
InChI Key PLRVIRHWKQKSAK-UHFFFAOYSA-N
Synonym 2,6-Dichloroisonicotinic Acid tert-Butyl Ester, tert-Butyl 2,6-Dichloro-4-pyridinecarboxylate, 2,6-Dichloro-4-pyridinecarboxylic Acid tert-Butyl Ester
PubChem CID 22594123
IUPAC Name tert-butyl 2,6-dichloropyridine-4-carboxylate
SMILES CC(C)(C)OC(=O)C1=CC(Cl)=NC(Cl)=C1

Specifications

Melting Point 82°C
Quantity 5 g
Formula Weight 248.10
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 2,6-Dichloroisonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.