Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl 2-aminoacetate | 6456-74-2 | 131.17 | 25 G
SDP

Catalog No. 5000449179
Encompass_Preferred

tert-Butyl 2-aminoacetate is a Glycine derivative that appears as a colorless to light yellow liquid. It is commercially used as an ergogenic supplement, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and helping prevent exercise-induced muscle damage.

  • Glycine derivative
  • Colorless to light yellow liquid
  • Purity of 99.64%
  • Store at 4°C, sealed, away from moisture and light
  • Store in solvent at -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-49179 Supplier Medchemexpress LLC Supplier No. 25GHYW007408
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl 2-aminoacetate is a Glycine derivative that appears as a colorless to light yellow liquid. It is commercially used as an ergogenic supplement, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and helping prevent exercise-induced muscle damage.

  • Glycine derivative
  • Colorless to light yellow liquid
  • Purity of 99.64%
  • Store at 4°C, sealed, away from moisture and light
  • Store in solvent at -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.