Learn More
Medchemexpress LLC tert-Butyl 2-aminoacetate | 6456-74-2 | 131.17 | 50 G
SDP
Catalog No. 5000460362
Encompass_Preferred
tert-Butyl 2-aminoacetate is a derivative of Glycine. Amino acids and their derivatives are widely recognized as beneficial ergogenic dietary substances, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress, and preventing exercise-induced muscle damage.
  • Derivative of Glycine
  • Used as ergogenic supplements
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • Recognized as beneficial ergogenic dietary substances
  • Reagent for peptide synthesis
  • Amino acid derivative
Catalog No. 50-004-60362 Supplier Medchemexpress LLC Supplier No. HYW00740850G
tert-Butyl 2-aminoacetate is a derivative of Glycine. Amino acids and their derivatives are widely recognized as beneficial ergogenic dietary substances, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress, and preventing exercise-induced muscle damage.
  • Derivative of Glycine
  • Used as ergogenic supplements
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • Recognized as beneficial ergogenic dietary substances
  • Reagent for peptide synthesis
  • Amino acid derivative