Learn More
Matrix Scientific tert-Butyl 2-aminoethylcarbamate, 57260-73-8, MFCD00191871, 25g
Supplier: Matrix Scientific 05373125G
Molecular Formula C7H16N2O2, Purity 95+%, Molecular Weight 160.22, Boiling Point 72/0.1mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.