Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 2-(hydroxymethyl)-1-indolinecarboxylate, 97%, Thermo Scientific™

Catalog No. MO01134DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO01134DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01134DE Supplier Thermo Scientific Chemicals Supplier No. MO01134DE

Chemical Identifiers

CAS 172647-87-9
Molecular Formula C14H19NO3
Molecular Weight (g/mol) 249.31
MDL Number MFCD06411549
InChI Key JNQNQSCDMKNYEX-UHFFFAOYSA-N
Synonym tert-butyl 2-hydroxymethyl-1-indolinecarboxylate, 1-t-butoxycarbonyl-2-hydroxymethylindoline, tert-butyl 2-hydroxymethyl-2,3-dihydroindole-1-carboxylate, 1-boc-2,3-dihydro-2-hydroxymethyl-1h-indole, tert-butyl 2-hydroxymethyl indoline-1-carboxylate, tert-butyl 2-hydroxymethyl indolinecarboxylate, tert-butyl 2-hydroxymethyl-2,3-dihydro-1h-indole-1-carboxylate, 1,1-dimethylethyl 2,3-dihydro-2 hydroxymethyl-1h-indole-1-carboxylate, 1,1-dimethylethyl 2,3-dihydro-2-hydroxymethyl-1h-indole-1-carboxylate
PubChem CID 2794669
IUPAC Name tert-butyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)CO

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 249.31
Percent Purity 97%
Chemical Name or Material tert-Butyl 2-(hydroxymethyl)-1-indolinecarboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.