Learn More
Medchemexpress LLC Tert-butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate | 535943-48-7 | MFCD24469377 | 96.0% | 286.41 g/mol | C12H18N2O2S2 | 100 MG
SDP
Catalog No. 5000284451
Encompass_Preferred
tert-Butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate is a Boc-protected carbamate containing a pyridin-2-yldisulfanyl group. It is commonly used as a reversible thiol-labeling reagent and as a protected intermediate in organic synthesis; the Boc (tert-butoxycarbonyl) group can be removed under mild acidic conditions to expose a primary amine for further derivatization. The pyridin-2-yldisulfanyl moiety enables disulfide exchange with cysteine residues or other thiols.
  • Reversible thiol labeling via disulfide exchange.
  • Boc-protected amine for controlled deprotection.
  • Suitable as a synthetic intermediate for derivatization.
  • Reported purity 96.0%.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-002-84451 Supplier Medchemexpress LLC Supplier No. HYW578278100MG
tert-Butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate is a Boc-protected carbamate containing a pyridin-2-yldisulfanyl group. It is commonly used as a reversible thiol-labeling reagent and as a protected intermediate in organic synthesis; the Boc (tert-butoxycarbonyl) group can be removed under mild acidic conditions to expose a primary amine for further derivatization. The pyridin-2-yldisulfanyl moiety enables disulfide exchange with cysteine residues or other thiols.
  • Reversible thiol labeling via disulfide exchange.
  • Boc-protected amine for controlled deprotection.
  • Suitable as a synthetic intermediate for derivatization.
  • Reported purity 96.0%.
  • Available in small pack sizes for laboratory use.