Learn More
Medchemexpress LLC Tert-butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate | 535943-48-7 | MFCD24469377 | 96.0% | 286.42 g/mol | C12H18N2O2S2 | 1 G
SDP
Catalog No. 5000284657
Encompass_Preferred
Tert-butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate is a Boc-protected carbamate reagent bearing a pyridinyl disulfide group. It is used as an intermediate in organic synthesis and for preparing thiol-reactive linkers where a protected aminoethyl spacer is required. Product specifications and handling instructions are provided in the supplier certificate of analysis.
  • Boc-protected carbamate useful for aminoethyl linker synthesis.
  • Pyridinyl disulfide moiety enables thiol-reactive conjugation.
  • High purity suitable for research applications and synthesis.
  • Stable under recommended storage: 4°C under nitrogen; in solvent, -80°C for long term.
  • Molecular formula C12H18N2O2S2 and molecular weight 286.42 g/mol.
Catalog No. 50-002-84657 Supplier Medchemexpress LLC Supplier No. HYW5782781G
Tert-butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate is a Boc-protected carbamate reagent bearing a pyridinyl disulfide group. It is used as an intermediate in organic synthesis and for preparing thiol-reactive linkers where a protected aminoethyl spacer is required. Product specifications and handling instructions are provided in the supplier certificate of analysis.
  • Boc-protected carbamate useful for aminoethyl linker synthesis.
  • Pyridinyl disulfide moiety enables thiol-reactive conjugation.
  • High purity suitable for research applications and synthesis.
  • Stable under recommended storage: 4°C under nitrogen; in solvent, -80°C for long term.
  • Molecular formula C12H18N2O2S2 and molecular weight 286.42 g/mol.