Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate | 535943-48-7 | MFCD24469377 | 96.0% | 286.42 g·mol⁻¹ | C12H18N2O2S2 | 500 MG
SDP

Catalog No. 5000283879
Encompass_Preferred

Tert-butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate is a carbamate derivative intended for research use. It is supplied as a white to off-white solid, with identity confirmed by 1H NMR and LCMS, and is commonly used as a synthetic building block or assay reagent. Handle using standard laboratory precautions and store under inert conditions to preserve stability.

  • White to off-white solid with identity confirmed by analytical data.
  • Purity ≥96.0% by NMR for reliable synthetic applications.
  • Molecular weight 286.42 g·mol⁻¹ and formula C12H18N2O2S2 for formulation calculations.
  • Storage under nitrogen at 4°C; in solution -80°C (6 months) or -20°C (1 month).
  • Supplied with batch-specific certificate of analysis and supporting spectra.

Catalog No. 50-002-83879 Supplier Medchemexpress LLC Supplier No. HYW578278500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tert-butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate is a carbamate derivative intended for research use. It is supplied as a white to off-white solid, with identity confirmed by 1H NMR and LCMS, and is commonly used as a synthetic building block or assay reagent. Handle using standard laboratory precautions and store under inert conditions to preserve stability.

  • White to off-white solid with identity confirmed by analytical data.
  • Purity ≥96.0% by NMR for reliable synthetic applications.
  • Molecular weight 286.42 g·mol⁻¹ and formula C12H18N2O2S2 for formulation calculations.
  • Storage under nitrogen at 4°C; in solution -80°C (6 months) or -20°C (1 month).
  • Supplied with batch-specific certificate of analysis and supporting spectra.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.