Learn More
Medchemexpress LLC Tert-butyl 3-(2-(2-(2-azidoethoxy)ethoxy)ethoxy)propanoate | 252881-73-5 | 303.3547 g/mol | C13H25N3O5 | 50 MG
SDP
Catalog No. 5000203676
Encompass_Preferred
N3-PEG3-CH2CH2-Boc is an azido-terminated PEG3 linker with a tert-butyl (Boc) protected carboxylic acid end, used as a cleavable PEG-based linker in antibody-drug conjugate and PROTAC synthesis, and as an azide reagent for click chemistry.
  • Azide-terminated PEG3 linker suitable for click chemistry.
  • tert-Butyl (Boc) protected carboxyl end for orthogonal deprotection.
  • Used in ADC and PROTAC linker assembly.
  • Available as a small pre-weighed laboratory pack for synthetic use.
Catalog No. 50-002-03676 Supplier Medchemexpress LLC Supplier No. HY4248950MG
N3-PEG3-CH2CH2-Boc is an azido-terminated PEG3 linker with a tert-butyl (Boc) protected carboxylic acid end, used as a cleavable PEG-based linker in antibody-drug conjugate and PROTAC synthesis, and as an azide reagent for click chemistry.
  • Azide-terminated PEG3 linker suitable for click chemistry.
  • tert-Butyl (Boc) protected carboxyl end for orthogonal deprotection.
  • Used in ADC and PROTAC linker assembly.
  • Available as a small pre-weighed laboratory pack for synthetic use.