Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate, 95%, Thermo Scientific™

Catalog No. AC451232500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451232500 250 mg
AC451230010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451232500 Supplier Thermo Scientific Chemicals Supplier No. 451232500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 898271-20-0
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
InChI Key XQNQFOKZUQWWDO-UHFFFAOYSA-N
Synonym 1-boc-3-aminoethylazetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, 1-boc-3-aminoethyl azetidine, 1-n-boc-3-aminoethyl azetidine, 1-boc-3-2-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate
PubChem CID 44630603
IUPAC Name tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)CCN

Specifications

Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 200.28
Percent Purity 95%
Chemical Name or Material tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.